Orders LinkContactUsLinkComponent
4-Bromo-2-fluorocinnamic acid
Glass bottle for solids, 25 g
  FLUO004225-25G
Marketsource
 :   FLUOROCHEM
4-Bromo-2-fluorocinnamic acid
  FLUO004225-25G
Marketsource
 :  149947-19-3
4-Bromo-2-fluorocinnamic acid
Glass bottle for solids, 25 g
 :  149947-19-3
179,00 €
1 * 25 g
  •  :149947-19-3
  •  :C₉H₆BrFO₂
  •  :MFCD00143267
  •  :245.05 g/mol
  •  :322 °C (1013 hPa)
  •  :219…223 °C
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g