Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Bromo-2-fluorocinnamic acid

4-Bromo-2-fluorocinnamic acid

Glass bottle for solids, 5 g
Catalog # FLUO004225-5G
Marketsource
Supplier:   FLUOROCHEM
4-Bromo-2-fluorocinnamic acid
Catalog # FLUO004225-5G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  149947-19-3
4-Bromo-2-fluorocinnamic acid

Glass bottle for solids, 5 g

CAS Number:  149947-19-3
Price
49,60 €
Log in for availability
1 * 5 g
Product Details & Documents
  • CAS Number:149947-19-3
  • Formula:C₉H₆BrFO₂
  • MDL Number:MFCD00143267
  • Molecular Weight:245.05 g/mol
  • Boiling Pt:322 °C (1013 hPa)
  • Melting Pt:219…223 °C
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.