Bismuth(III) fluoride
Glass bottle for solids, 500 g
Catalog # FLUO004113-500G
Marketsource
Supplier: FLUOROCHEM
Bismuth(III) fluoride
Catalog # FLUO004113-500G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 7787-61-3
Jump to:
Product Details & Documents
- CAS Number:7787-61-3
- Formula:BiF₃
- MDL Number:MFCD00010893
- Molecular Weight:265.98 g/mol
- Merck Index:12,01305
- Boiling Pt:230 °C (1013 hPa)
- Melting Pt:150.9…151.9 °C
- Density:3.7 g/cm³ (160 °C)
- EINECS:232-124-8
- ADR:8,III
- :Density: 3.7 g/cm³ (160 °C)
- UN:3260
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



