Order Entry
Germany
Orders LinkContactUsLinkComponent
Bismuth(III) fluoride

Bismuth(III) fluoride

Glass bottle for solids, 100 g
Catalog # FLUO004113-100G
Corrosive
Marketsource
Supplier:   FLUOROCHEM
Bismuth(III) fluoride
Catalog # FLUO004113-100G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  7787-61-3
Bismuth(III) fluoride

Glass bottle for solids, 100 g

CAS Number:  7787-61-3
Price
492,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:7787-61-3
  • Formula:BiF₃
  • MDL Number:MFCD00010893
  • Molecular Weight:265.98 g/mol
  • Merck Index:12,01305
  • Boiling Pt:230 °C (1013 hPa)
  • Melting Pt:150.9…151.9 °C
  • Density:3.7 g/cm³ (160 °C)
  • EINECS:232-124-8
  • ADR:8,III
  •  :Density: 3.7 g/cm³ (160 °C)
  • UN:3260
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.