Order Entry
Germany
Orders LinkContactUsLinkComponent
2-Ethoxythiazole

2-Ethoxythiazole

Glass bottle for solids, 25 g
Catalog # FLUO002763-25G
Marketsource
Supplier:   FLUOROCHEM
2-Ethoxythiazole
Catalog # FLUO002763-25G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  15679-19-3
2-Ethoxythiazole

Glass bottle for solids, 25 g

CAS Number:  15679-19-3
Price
95,40 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:15679-19-3
  • Formula:C₅H₇NOS
  • MDL Number:MFCD00055026
  • Molecular Weight:129.18 g/mol
  • EINECS:239-760-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.