Order Entry
Germany
Orders LinkContactUsLinkComponent
6-Bromoquinaldine

6-Bromoquinaldine

Glass bottle for solids, 25 g
Catalog # FLUO002521-25G
Marketsource
Supplier:   FLUOROCHEM
6-Bromoquinaldine
Catalog # FLUO002521-25G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  877-42-9
6-Bromoquinaldine

Glass bottle for solids, 25 g

Wechseln zu:
CAS Number:  877-42-9
Price
156,00 €
Anmelden für die Verfügbarkeit
1 * 25 g
Product Details & Documents
  • CAS-Nummer:877-42-9
  • Formel::C₁₀H₈BrN
  • MDL-Nummer:MFCD00079724
  • Molekulargewicht:222.08 g/mol
Spezifikationen:

Spezifikationen:

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Spezifikationen:

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.