3,4,5-Trifluorophenol
10 g
Catalog # FLUO002074-10G
Marketsource
Supplier: FLUOROCHEM
3,4,5-Trifluorophenol
Catalog # FLUO002074-10G
Supplier: FLUOROCHEM
Marketsource
CAS Number: 99627-05-1
Jump to:
Product Details & Documents
- CAS Number:99627-05-1
- Formula:C₆H₃F₃O
- MDL Number:MFCD00042427
- Molecular Weight:148.08 g/mol
- Density:1.629 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:436-120-9
- UN:3261
- ADR:8,II
- :Density: 1.629 g/cm³ (20 °C)
Specifications
Specifications
- Pk:10 g
- Cat. No.:FLUO002074-10G



