Order Entry
Germany
Orders LinkContactUsLinkComponent
 

3,4,5-Trifluorophenol

Glass bottle for solids, 1 g
Catalog # FLUO002074-1G
Marketsource
Supplier:   FLUOROCHEM
3,4,5-Trifluorophenol
Catalog # FLUO002074-1G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  99627-05-1
3,4,5-Trifluorophenol

Glass bottle for solids, 1 g

CAS Number:  99627-05-1
Price
31,50 €
Log in for availability
1 * 1 g
Product Details & Documents
  • CAS Number:99627-05-1
  • Formula:C₆H₃F₃O
  • MDL Number:MFCD00042427
  • Molecular Weight:148.08 g/mol
  • Density:1.629 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:436-120-9
  • UN:3261
  • ADR:8,II
  •  :Density: 1.629 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications