2,4-Difluorphenol
Glasflasche für Feststoffe, 250 g
Catalog # FLUO001926-250G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
- CAS Number:367-27-1
- Formula:C₆H₄F₂O
- MDL Number:MFCD00009715
- Molecular Weight:130.09 g/mol
- Boiling Pt:146 °C (1013 hPa)
- Melting Pt:20 °C
- Density:1,36 g/cm³ (20 °C)
- EINECS:206-688-0
- UN:2921
- ADR:8,II
Specifications
Specifications
- Packung:Glasflasche für Feststoffe
- VE:250 g



