Order Entry
Germany
Orders LinkContactUsLinkComponent
2,4-Difluorophenol

2,4-Difluorophenol

Glass bottle for solids, 25 g
Catalog # FLUO001926-25G
Marketsource
Supplier:   FLUOROCHEM
2,4-Difluorophenol
Catalog # FLUO001926-25G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  367-27-1
2,4-Difluorophenol

Glass bottle for solids, 25 g

CAS Number:  367-27-1
Price
57,10 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:367-27-1
  • Formula:C₆H₄F₂O
  • MDL Number:MFCD00009715
  • Molecular Weight:130.09 g/mol
  • Boiling Pt:146 °C (1013 hPa)
  • Melting Pt:20 °C
  • Density:1.36 g/cm³ (20 °C)
  • EINECS:206-688-0
  • UN:2921
  • ADR:8,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.