Order Entry
Germany
Orders LinkContactUsLinkComponent
 

3,5-Difluorophenol

Glass bottle for solids, 25 g
Catalog # FLUO001659-25G
Marketsource
Supplier:   FLUOROCHEM
3,5-Difluorophenol
Catalog # FLUO001659-25G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  2713-34-0
3,5-Difluorophenol

Glass bottle for solids, 25 g

CAS Number:  2713-34-0
Price
83,50 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:2713-34-0
  • Formula:C₆H₄F₂O
  • MDL Number:MFCD00002255
  • Molecular Weight:130.09 g/mol
  • Boiling Pt:65…68 °C (15 mmHg)
  • Melting Pt:55 °C
  • Density:1.566 g/cm³ (20 °C)
  • UN:3261
  • ADR:8,II
  •  :Density: 1.566 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications