2'-Fluoroacetophenone
Glass bottle for solids, 100 g
FLUO001616-100G
Marketsource
: 1-(2-Fluorophenyl)ethanone, o-Fluoroacetophenone
- :445-27-2
- :C₈H₇FO
- :MFCD00000320
- :138.14 g/mol
- :82…83 °C (13,3 hPa)
- :82…83 °C (13.3 hPa)
- :1,12 g/cm³ (20 °C)
- :Ambient
- :207-156-0
- Pack type:Glass bottle for solids
- Pk:100 g