Order Entry
Germany
Orders LinkContactUsLinkComponent
4-Fluorophenol

4-Fluorophenol

Glass bottle for solids, 100 g
Catalog # FLUO001291-100G
Irritant
Marketsource
Supplier:   FLUOROCHEM
4-Fluorophenol
Catalog # FLUO001291-100G
Supplier:  FLUOROCHEM
Marketsource
CAS Number:  371-41-5
4-Fluorophenol

Glass bottle for solids, 100 g

CAS Number:  371-41-5
Price
95,40 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
  • CAS Number:371-41-5
  • Formula:C₆H₅FO
  • MDL Number:MFCD00002316
  • Molecular Weight:112.1 g/mol
  • Boiling Pt:185 °C (1013 hPa)
  • Melting Pt:44…47 °C
  • Density:1.22 g/cm³ (20 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Ambient
  • EINECS:206-736-0
  • UN:2923
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.