Order Entry
Germany
Orders LinkContactUsLinkComponent
Phenoxyacetamide

Phenoxyacetamide

Glass bottle for solids, 25 g
Artikelnummer FLUO001028-25G
Marketsource
Hersteller:   FLUOROCHEM
Phenoxyacetamide
Artikelnummer FLUO001028-25G
Hersteller:  FLUOROCHEM
Marketsource
CAS-Nummer:  621-88-5
Phenoxyacetamide

Glass bottle for solids, 25 g

CAS-Nummer:  621-88-5
Preis
951,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-Phenoxyacetamide
  • CAS Number:621-88-5
  • Formula:C₈H₉NO₂
  • MDL Number:MFCD00017140
  • Molecular Weight:151.16 g/mol
  • EINECS:210-713-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications