3'-Nitroformanilide
Glass bottle for solids, 250 g
Catalog # MENAIP63.250
Supplier: MENAI
Jump to:
Product Details & Documents
Synonyms: 3-Nitroformanilide
- CAS Number:102-38-5
- Formula:C₇H₆N₂O₃
- MDL Number:MFCD00017014
- Molecular Weight:166.14 g/mol
- Melting Pt:134 °C
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



