Methyl-2-formylbenzoate
Glass bottle for solids, 1 g
Catalog # MAYBTL00698DA
Marketsource
Supplier: Thermo Scientific
Methyl-2-formylbenzoate
Catalog # MAYBTL00698DA
Supplier: Thermo Scientific
Marketsource
CAS Number: 4122-56-9
Jump to:
Product Details & Documents
Synonyms: Methyl 2-formylbenzoate
- CAS Number:4122-56-9
- Formula:C₉H₈O₃
- MDL Number:MFCD00204231
- Molecular Weight:164.1580 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



