4-Hydroxybenzhydrazide
Glass bottle for solids, 1 g
MAYBSEW04591.1
Marketsource
: 4-Hydroxybenzoic hydrazide, 4-Hydroxybenzohydrazide, p-Hydroxybenzoic acid hydrazide, 4-Hydroxybenzene-1-carbohydrazide, 4-Hydroxybenzoic acid hydrazide
- :5351-23-5
- :C₇H₈N₂O₂
- :MFCD00007605
- :152.15 g/mol
- :370 °C (1013 hPa)
- :266…269 °C
- :Ambient
- :226-326-5
- Pack type:Glass bottle for solids
- Pk:1 g