Orders LinkContactUsLinkComponent
Cyclooctanone
Glass bottle for solids, 50 g
  MAYBSB01231.50
Marketsource
Cyclooctanone
  MAYBSB01231.50
Marketsource
 :  502-49-8
Cyclooctanone
Glass bottle for solids, 50 g
 :  502-49-8
1 * 50 g
  •  :502-49-8
  •  :C₈H₁₄O
  •  :MFCD00001754
  •  :126.2 g/mol
  •  :196 °C (1013 hPa)
  •  :39…41 °C
  •  :0,9581 g/cm³ (20 °C)
  •  :Ambient
  •  :207-940-2
  •  :1759
  •  :8,III
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g