Cyclooctanone
Glass bottle for solids, 10 g
Catalog # MAYBSB01231.10
Marketsource
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:502-49-8
- Formula:C₈H₁₄O
- MDL Number:MFCD00001754
- Molecular Weight:126.2 g/mol
- Boiling Pt:196 °C (1013 hPa)
- Melting Pt:39…41 °C
- Density:0,9581 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:207-940-2
- UN:1759
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



