Diethyl ethyl(phenyl)malonate
Glass bottle for solids, 50 g
Catalog # MAYBSB00827.50
Marketsource
Supplier: Thermo Scientific
Diethyl ethyl(phenyl)malonate
Catalog # MAYBSB00827.50
Supplier: Thermo Scientific
Marketsource
CAS Number: 76-67-5
Jump to:
Product Details & Documents
Synonyms: Diethyl 2-ethyl-2-phenylmalonate
- CAS Number:76-67-5
- Formula:C₁₅H₂₀O₄
- MDL Number:MFCD00040759
- Molecular Weight:264.32 g/mol
- EINECS:200-978-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



