Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Diethyl ethyl(phenyl)malonate

Glass bottle for solids, 10 g
Catalog # MAYBSB00827.10
Marketsource
Supplier:   Thermo Scientific
Diethyl ethyl(phenyl)malonate
Catalog # MAYBSB00827.10
Supplier:  Thermo Scientific
Marketsource
CAS Number:  76-67-5
Diethyl ethyl(phenyl)malonate

Glass bottle for solids, 10 g

CAS Number:  76-67-5
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Diethyl 2-ethyl-2-phenylmalonate
  • CAS Number:76-67-5
  • Formula:C₁₅H₂₀O₄
  • MDL Number:MFCD00040759
  • Molecular Weight:264.32 g/mol
  • EINECS:200-978-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications