4-Fluorophenol
Glass bottle for solids, 50 g
Catalog # MAYBSB00777.50
Marketsource
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: p-Fluorophenol, 1-Fluoro-4-hydroxybenzene
- CAS Number:371-41-5
- Formula:C₆H₅FO
- MDL Number:MFCD00002316
- Molecular Weight:112.1 g/mol
- Boiling Pt:185 °C (1013 hPa)
- Melting Pt:44…47 °C
- Density:1.22 g/cm³ (20 °C)
- Flash Pt:68 °C
- Storage Temperature:Ambient
- EINECS:206-736-0
- UN:2923
- ADR:8,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



