2,4,6-Trimethylaniline
Glass bottle for solids, 1 g
Catalog # MAYBSB00547.1
Marketsource
Supplier: Thermo Scientific
2,4,6-Trimethylaniline
Catalog # MAYBSB00547.1
Supplier: Thermo Scientific
Marketsource
CAS Number: 88-05-1
Jump to:
Product Details & Documents
- CAS Number:88-05-1
- Formula:C₉H₁₃N
- MDL Number:MFCD00007740
- Molecular Weight:135.21 g/mol
- Boiling Pt:233 °C (1013 hPa)
- Melting Pt:–5 °C
- Density:0,96 g/cm³ (20 °C)
- Flash Pt:96 °C
- Storage Temperature:Ambient
- EINECS:201-794-3
- UN:2810
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



