2,4,6-Trimethylaniline
Glass bottle for solids, 250 mg
CHMPSB00547.250
: CHEMPUR
- :88-05-1
- :C₉H₁₃N
- :MFCD00007740
- :135.21 g/mol
- :233 °C (1013 hPa)
- :–5 °C
- :0,96 g/cm³ (20 °C)
- :96 °C
- :Ambient
- :201-794-3
- :2810
- :6.1,III
- Pack type:Glass bottle for solids
- Pk:250 mg