5-Iodoindole
Glass bottle for solids, 1 g
Catalog # MAYBRH00726.1
Marketsource
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Synonyms: 5-Jodindol, 5-Jod-1H-indol
- CAS Number:16066-91-4
- Formula:C₈H₆IN
- MDL Number:MFCD00220065
- Molecular Weight:243.05 g/mol
Specifications
Specifications
- Packung:Glass bottle for solids
- VE:1 g



