5-Iodoindole
Glass bottle for solids, 250 mg
Catalog # CHMPRH00726.250
Supplier: CHEMPUR
Jump to:
Product Details & Documents
- CAS Number:16066-91-4
- Formula:C₈H₆IN
- MDL Number:MFCD00220065
- Molecular Weight:243.05 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg



