Orders LinkContactUsLinkComponent
Electrode Cables
Electrode Cables
Electrode cable, for connecting electrodes with Metrohm plug-in head G to Metrohm instruments (socket F), 2 m
Catalog # METR6.2104.030
Supplier:   METROHM
Electrode Cables
Catalog # METR6.2104.030
Supplier:  METROHM
Electrode Cables
Electrode cable, for connecting electrodes with Metrohm plug-in head G to Metrohm instruments (socket F), 2 m
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Electrode cable, for connecting electrodes with Metrohm plug-in head G to Metrohm instruments (socket F)
  • Length:
    2 m
  • Seq:
    02