Ethoxytrimethylsilane
Glass bottle for solids, 100 g
Catalog # SILNSILS18600.100
Marketsource
Supplier: CHEMPUR SILANE
Ethoxytrimethylsilane
Catalog # SILNSILS18600.100
Supplier: CHEMPUR SILANE
Marketsource
CAS Number: 1825-62-3
Jump to:
Product Details & Documents
Synonyms: Trimethyl(ethoxy)silane
- CAS Number:1825-62-3
- Formula:C₅H₁₄OSi
- MDL Number:MFCD00009069
- Molecular Weight:118.25 g/mol
- Boiling Pt:75 °C (1013 hPa)
- Melting Pt:–83 °C
- Density:0,7573 g/cm³ (20 °C)
- EINECS:217-370-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



