Ethoxytrimethylsilane
Glass bottle for solids, 25 g
SILNSILS18600.25
Marketsource
: Trimethyl(ethoxy)silane
- :1825-62-3
- :C₅H₁₄OSi
- :MFCD00009069
- :118.25 g/mol
- :75 °C (1013 hPa)
- :–83 °C
- :0,7573 g/cm³ (20 °C)
- :217-370-6
- Pack type:Glass bottle for solids
- Pk:25 g