2-Brom-4'-pentyloacetophenone
Glass bottle for solids, 1 g
Catalog # MAYBBTB12841DA
Marketsource
Supplier: Thermo Scientific
2-Brom-4'-pentyloacetophenone
Catalog # MAYBBTB12841DA
Supplier: Thermo Scientific
Marketsource
CAS Number: 64328-68-3
Jump to:
Product Details & Documents
Synonyms: 4-(Pent-1-yl)phenacyl bromide
- CAS Number:64328-68-3
- Formula:C₁₃H₁₇BrO
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



