2-(Methylthio)nicotinoyl chloride
Glass bottle for solids, 10 g
Catalog # MAYBBTB06290.10
Marketsource
Supplier: Thermo Scientific
2-(Methylthio)nicotinoyl chloride
Catalog # MAYBBTB06290.10
Supplier: Thermo Scientific
Marketsource
CAS Number: 97936-43-1
Jump to:
Product Details & Documents
- CAS Number:97936-43-1
- Formula:C₇H₆ClNOS
- MDL Number:MFCD00051678
- Molecular Weight:187.65 g/mol
- UN:3261
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



