5'-Bromo-2'-hydroxyacetophenone
1 g
Catalog # MAYBBTB04266.1
Marketsource
Supplier: Thermo Scientific
5'-Bromo-2'-hydroxyacetophenone
Catalog # MAYBBTB04266.1
Supplier: Thermo Scientific
Marketsource
CAS Number: 1450-75-5
Jump to:
Product Details & Documents
Synonyms: 1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone
- CAS Number:1450-75-5
- Formula:C₈H₇BrO₂
- MDL Number:MFCD00191850
- Molecular Weight:215.05
Specifications
Specifications
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



