5'-Bromo-2'-hydroxyacetophenone
250 mg
Catalog # CHMPBTB04266.250
Supplier: CHEMPUR
Jump to:
Product Details & Documents
Synonyms: 1-(5-Bromo-2-hydroxyphenyl)ethan-1-one, 1-(5-Bromo-2-hydroxyphenyl)ethanone
- CAS Number:1450-75-5
- Formula:C₈H₇BrO₂
- MDL Number:MFCD00191850
- Molecular Weight:215.05
Specifications
Specifications
- Pk:250 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



