2-(2,4-Difluorophenoxy)-3-nitropyridine
Glass bottle for solids, 10 g
MAYBBTB08013.10
Marketsource
2-(2,4-Difluorophenoxy)-3-nitropyridine
MAYBBTB08013.10
Marketsource
: 175135-62-3
- :175135-62-3
- :C₁₁H₆F₂N₂O₃
- :MFCD00067800
- :252.18 g/mol
- Pack type:Glass bottle for solids
- Pk:10 g