ω,4-Dibromoacetophenone
Glass bottle for solids, 50 g
Catalog # MAYBBTB07599.50
Marketsource
Supplier: Thermo Scientific
ω,4-Dibromoacetophenone
Catalog # MAYBBTB07599.50
Supplier: Thermo Scientific
Marketsource
CAS Number: 99-73-0
Wechseln zu:
Product Details & Documents
Synonyme: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
- CAS-Nummer:99-73-0
- Formel::C₈H₆Br₂O
- MDL-Nummer:MFCD00000200
- Molekulargewicht:277.94 g/mol
- Merck-Index:13,01412
- Siedepunkt:310 °C (1013 hPa)
- Schmelzpunkt:107…108 °C
- Lagertemperatur:Refrigerator
- EINECS:202-783-6
- UN:3261
- ADR:8,II
Spezifikationen:
Spezifikationen:
- Pack type:Glass bottle for solids
- Pk:50 g
Spezifikationen:
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



