ω,4-Dibromoacetophenone
Glass bottle for solids, 1 g
Catalog # MAYBBTB07599.1
Marketsource
Supplier: Thermo Scientific
ω,4-Dibromoacetophenone
Catalog # MAYBBTB07599.1
Supplier: Thermo Scientific
Marketsource
CAS Number: 99-73-0
Jump to:
Product Details & Documents
Synonyms: Tau Protein Kinase I Inhibitor, 2,4'-Dibromoacetophenone, Glycogen Synthase Kinase-3β Inhibitor VII, α,4-Dibromoacetophenone, GSK-3β Inhibitor VII, TPK I Inhibitor, 2-Bromo-1-(4-bromophenyl)-1-ethanone, 4-Bromophenacyl bromide
- CAS Number:99-73-0
- Formula:C₈H₆Br₂O
- MDL Number:MFCD00000200
- Molecular Weight:277.94 g/mol
- Merck Index:13,01412
- Boiling Pt:310 °C (1013 hPa)
- Melting Pt:107…108 °C
- Storage Temperature:Refrigerator
- EINECS:202-783-6
- UN:3261
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



