N-(2-Hydroxyethyl)-2-phenylacetamide
Glass bottle for solids, 1 g
Catalog # BIOT8F-926.1
Marketsource
Supplier: BIONET RESEARCH
N-(2-Hydroxyethyl)-2-phenylacetamide
Catalog # BIOT8F-926.1
Supplier: BIONET RESEARCH
Marketsource
CAS Number: 6269-99-4
Jump to:
Product Details & Documents
- CAS Number:6269-99-4
- Formula:C₁₀H₁₃NO₂
- MDL Number:MFCD00043506
- EINECS:228-444-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



