N-(2-Nitrobenzoyl)morpholine
Glass bottle for solids, 500 mg
Catalog # BIOT7X-5058.500
Marketsource
Supplier: BIONET RESEARCH
N-(2-Nitrobenzoyl)morpholine
Catalog # BIOT7X-5058.500
Supplier: BIONET RESEARCH
Marketsource
CAS Number: 26162-89-0
Jump to:
Product Details & Documents
Synonyms: Morpholino(2-nitrophenyl)methanone
- CAS Number:26162-89-0
- Formula:C₁₁H₁₂N₂O₄
- MDL Number:MFCD00586005
- Molecular Weight:236.23 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



