2-Hydroxy-3'-nitroacetophenone
10 g
Catalog # BIOT3X-0728.10
Marketsource
Supplier: BIONET RESEARCH
2-Hydroxy-3'-nitroacetophenone
Catalog # BIOT3X-0728.10
Supplier: BIONET RESEARCH
Marketsource
CAS Number: 72802-41-6
Jump to:
Product Details & Documents
Synonyms: α-Hydroxy-3'-nitroacetophenone, 2-Hydroxy-1-(3-nitrophenyl)-1-ethanone
- CAS Number:72802-41-6
- Formula:C₈H₇NO₄
- MDL Number:MFCD04973723
- Molecular Weight:181.15
Specifications
Specifications
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



