2-(4-Chlorophenoxy)benzaldehyde
Glass bottle for solids, 1 g
Catalog # BIOT11N-050.1
Marketsource
Supplier: BIONET RESEARCH
2-(4-Chlorophenoxy)benzaldehyde
Catalog # BIOT11N-050.1
Supplier: BIONET RESEARCH
Marketsource
CAS Number: 111826-11-0
Jump to:
Product Details & Documents
- CAS Number:111826-11-0
- Formula:C₁₃H₉ClO₂
- MDL Number:MFCD01568734
- Molecular Weight:232.6600 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



