2-(4-Chlorophenoxy)benzaldehyde
Glass bottle for solids, 500 mg
Catalog # BIOT11N-050.500
Marketsource
Supplier: BIONET RESEARCH
2-(4-Chlorophenoxy)benzaldehyde
Catalog # BIOT11N-050.500
Supplier: BIONET RESEARCH
Marketsource
CAS Number: 111826-11-0
Jump to:
Product Details & Documents
- CAS Number:111826-11-0
- Formula:C₁₃H₉ClO₂
- MDL Number:MFCD01568734
- Molecular Weight:232.6600 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



