2-(4-Fluorophenoxy)benzaldehyde
Glass bottle for solids, 500 mg
Catalog # BIOT12N-071.500
Marketsource
Supplier: BIONET RESEARCH
2-(4-Fluorophenoxy)benzaldehyde
Catalog # BIOT12N-071.500
Supplier: BIONET RESEARCH
Marketsource
CAS Number: 320423-61-8
Jump to:
Product Details & Documents
- CAS Number:320423-61-8
- Formula:C₁₃H₉FO₂
- MDL Number:MFCD01568770
- Molecular Weight:216.21 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg



