2-(Ammoniooxy)propane chloride
Glass bottle for solids, 10 g
Catalog # BIOT12H-943.10
Marketsource
Supplier: BIONET RESEARCH
2-(Ammoniooxy)propane chloride
Catalog # BIOT12H-943.10
Supplier: BIONET RESEARCH
Marketsource
CAS Number: 4490-81-7
Jump to:
Product Details & Documents
Synonyms: O-propan-2-ylhydroxylamine hydrochloride, O-Isopropylhydroxylamine hydrochloride
- CAS Number:4490-81-7
- Formula:C₃H₁₀ClNO
- MDL Number:MFCD00231457
- Molecular Weight:111.5706 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



