2-(Ammoniooxy)propane chloride
Glass bottle for solids, 1 g
Catalog # BIOT12H-943.1
Marketsource
Supplier: BIONET RESEARCH
2-(Ammoniooxy)propane chloride
Catalog # BIOT12H-943.1
Supplier: BIONET RESEARCH
Marketsource
CAS Number: 4490-81-7
Jump to:
Product Details & Documents
Synonyms: O-propan-2-ylhydroxylamine hydrochloride, O-Isopropylhydroxylamine hydrochloride
- CAS Number:4490-81-7
- Formula:C₃H₁₀ClNO
- MDL Number:MFCD00231457
- Molecular Weight:111.5706 g/mol
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



