Buffer, SSC solution 20X concentrate (saline-sodium citrate)
SSC buffer solution 20X concentrate (saline-sodium citrate), Glass bottle, 1 L
Catalog # AMBI9763
Marketsource
Supplier: Ambion
Buffer, SSC solution 20X concentrate (saline-sodium citrate)
Catalog # AMBI9763
Supplier: Ambion
Marketsource
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Description:SSC buffer solution 20X concentrate (saline-sodium citrate)



