Order Entry
Germany
Orders LinkContactUsLinkComponent
SlideHyb Buffer #1 1 * 10 ml

SlideHyb Buffer #1 1 * 10 ml

Catalog # AMBI8861
Marketsource
Supplier:   Ambion
SlideHyb Buffer #1 1 * 10 ml
Catalog # AMBI8861
Supplier:  Ambion
Marketsource
SlideHyb Buffer #1 1 * 10 ml
Price
Inquire For Price
Log in for availability
1 * 10 mL
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.