ICP standard silver 1.000 g Ag/l AgNO3 in nitric acid 0.5 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Catalog # KRAF03957.0700
Supplier: Bernd Kraft
ICP standard silver 1.000 g Ag/l AgNO3 in nitric acid 0.5 mol/l traceable to NIST Content: 0,1 l 1 * 100 mL
Catalog # KRAF03957.0700
Supplier: Bernd Kraft
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
Supplier: Merck
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR®, Chlorate 1,000 mg/L in Water standard for ion chromatography
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
UV/Visible standards, Supelco®
Supplier: Merck