Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
D-ASPARAGINE MONOHYDRATE 1 * 500 g
Catalog # ICNA0210078590
Supplier:
MP Biomedicals
Expand image
D-ASPARAGINE MONOHYDRATE 1 * 500 g
Catalog # ICNA0210078590
Supplier:
MP Biomedicals