0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
ISONICOTINIC ACID 1 * 100 g
ICNA210209580
:
MP Biomedicals
ISONICOTINIC ACID 1 * 100 g
ICNA210209580
:
MP Biomedicals