Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SMA I5' . . CCC/GGG . . 3' 1 * 2 KU
Catalog # ICNA215205902
Supplier:
MP Biomedicals
Expand image
SMA I5' . . CCC/GGG . . 3' 1 * 2 KU
Catalog # ICNA215205902
Supplier:
MP Biomedicals